
N-Methylhydroxylammonium chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4229-44-1
:
Formula:CH₆ClNO
MDL Number:MFCD00012597
:83.52 g/mol
Melting Pt:80…86 °C
EINECS:224-181-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together