5-Chloro-3-methylbenzo[b]thiophene
Catalog # APOSOR1242-10G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:182.67 g/mol
Storage Temperature:Ambient
CAS Number:19404-18-3
Formula:C₉H₇ClS
MDL Number:MFCD00052506