5-Bromo-4-methyl-1H-imidazole
Catalog # APOSOR1225-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:161.0000 g/mol
Melting Pt:140…143 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:15813-08-8
Formula:C₄H₅BrN₂
MDL Number:MFCD06659898