Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromoethyl-4-(tert-butyl)phenyl ether
2-Bromoethyl-4-(tert-butyl)phenyl ether
2-Bromoethyl-4-(tert-butyl)phenyl ether
APOSOR12040-1G
:
Apollo Scientific
:
2-Bromoethyl-4-(tert-butyl)phenyl ether
APOSOR12040-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
257.18 g/mol
:
95 °C/0.38mmHg
:
GHS07
:
Ambient
:
5952-59-0
:
C₁₂H₁₇BrO
:
MFCD02030708