Order Entry
United Kingdom
ContactUsLinkComponent
[4-(4-Chlorophenyl)phenyl]sulphonyl chloride 96%
Corrosive
[4-(4-Chlorophenyl)phenyl]sulphonyl chloride 96%
Catalog # APOSOR11342-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
[4-(4-Chlorophenyl)phenyl]sulphonyl chloride 96%
Catalog # APOSOR11342-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :287.17 g/mol
Melting Pt:108…110 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05
CAS Number:20443-74-7
Formula:C₁₂H₈Cl₂O₂S
MDL Number:MFCD01631918