Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
ω-Bromo-3-nitroacetophenone

ω-Bromo-3-nitroacetophenone

Bottle
Catalog # APOSOR11094-500G
Corrosive
New Product
Supplier:   Apollo Scientific
ω-Bromo-3-nitroacetophenone
Catalog # APOSOR11094-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  2227-64-7
ω-Bromo-3-nitroacetophenone

Bottle

CAS Number:  2227-64-7
Price
£146.00
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  3-Nitrophenacyl bromide, 2-Bromo-3'-nitroacetophenone, α-Bromo-3-nitroacetophenone, Bromomethyl m-nitrophenyl ketone
  • CAS Number:2227-64-7
  • Formula:C₈H₆BrNO₃
  • MDL Number:MFCD00024512
  • Molecular Weight:244.04 g/mol
  • Melting Pt:89…96 °C
  • Storage Temperature:Ambient
  • EINECS:218-764-0
  • UN:3261
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.