
1,3,5-Trimethyl-1H-pyrazole-4-carbonyl chloride
Catalog # APOSOR110409-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:172.62 g/mol
Boiling Pt:140…150 °C/12mmHg
Melting Pt:67…68 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
CAS Number:98298-63-6
Formula:C₇H₉ClN₂O
MDL Number:MFCD03421589