

2-Chloropyrazine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14508-49-7
:
Formula:C₄H₃ClN₂
MDL Number:MFCD00006124
:114.53 g/mol
Boiling Pt:153…154 °C
Density:1.283 g/cm³ (25 °C)
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:238-517-0
UN:1993
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together