
5-Chloro-2-nitrobenzaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6628-86-0
:
Formula:C₇H₄ClNO₃
MDL Number:MFCD00007289
Melting Pt:76…78 °C
:185.57 g/mol
EINECS:229-614-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together