Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2-methylanisole
4-Chloro-2-methylanisole
4-Chloro-2-methylanisole
APOSOR1079-25G
:
Apollo Scientific
:
4-Chloro-2-methylanisole
APOSOR1079-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
156.61 g/mol
:
Ambient
:
3260-85-3
:
C₈H₉ClO
:
MFCD00070771