
4,5-Dichlorophthalonitrile 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:139152-08-2
:
Formula:C₈H₂Cl₂N₂
MDL Number:MFCD00191408
:197.02 g/mol
Melting Pt:180…184 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together