Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Benzyldimethylammonio)propanesulphonate
3-(Benzyldimethylammonio)propanesulphonate 95+%
3-(Benzyldimethylammonio)propanesulphonate 95+%
APOSOR1004-1G
:
Apollo Scientific
:
3-(Benzyldimethylammonio)propanesulphonate 95+%
APOSOR1004-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
257.348 g/mol
:
245…248 °C
:
GHS07
:
Ambient
:
81239-45-4
:
C₁₂H₁₉NO₃S
:
MFCD00225018