
(3,4-Dichlorophenyl)acetic acid methyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6725-44-6
:
Formula:(C₆H₃Cl₂)CH₂COOCH₃
MDL Number:MFCD00800680
:219.07 g/mol
Melting Pt:25…27 °C
Density:1.33 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together