4-Phenoxy-1H-indazol-3-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:816454-31-6
:
Formula:C₁₃H₁₁N₃O
MDL Number:MFCD09864800
:225.25 g/mol
Melting Pt:96…98 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together