

tert-Butylchlorodimethylsilane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18162-48-6
:
Formula:C₆H₁₅ClSi
MDL Number:MFCD00000501
:150.72 g/mol
Boiling Pt:124…126 °C
Melting Pt:86…89 °C
Density:0.81 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:242-042-4
UN:2925
ADR:4.1+8,II
:GHS02
:GHS05
Frequently Bought Together