
1,2,3-Benzothiadiazole-5-carbonyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:321309-32-4
:
Formula:C₇H₃ClN₂OS
MDL Number:MFCD01764004
:198.63 g/mol
Melting Pt:93…96 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together