
1-Benzhydrylazetidine-3-carboxylic acid 95+%
Catalog # APOSOR01635-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:267.33 g/mol
Melting Pt:198 °C (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:36476-87-6
:MFCD01566539
Formula:C₁₇H₁₇NO₂
MDL Number:MFCD01566538