Order Entry
United Kingdom
ContactUsLinkComponent
 
Vanadium(V) oxide 99.99+%
Catalog # APOSIN3778-10G
Supplier:  Apollo Scientific
CAS Number:  
Vanadium(V) oxide 99.99+%
Catalog # APOSIN3778-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
 :
 :181.88 g/mol
EINECS:215-239-8
UN:2862
 :GHS08
 :GHS05
 :GHS09
 :GHS07
Storage Temperature:Ambient
CAS Number:1314-62-1
Formula:O₅V₂
MDL Number:MFCD00011457
Boiling Pt:1750 °C
Melting Pt:690 °C
Density:3.357 g/cm³ (25 °C)
ADR:6.1 III
Merck Index:13,09987