Order Entry
United Kingdom
ContactUsLinkComponent
Terbium(III) acetylacetonate 99,9%
Terbium(III) acetylacetonate 99,9%
Catalog # APOSIN3490-2G
Supplier:  Apollo Scientific
CAS Number:  
Terbium(III) acetylacetonate 99,9%
Catalog # APOSIN3490-2G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    2 g
  • Seq:
    3107

Specifications

About this item

 :
 :456.25 g/mol
Storage Temperature:Ambient
CAS Number:14284-95-8
Formula:C₁₅H₂₁O₆Tb
MDL Number:MFCD00013503