
Dipotassium tetraborate tetrahydrate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:12045-78-2
:
Formula:B₄K₂O₇·4H₂O
MDL Number:MFCD00149924
:305.49 g/mol
EINECS:215-575-5
:GHS08
Storage Temperature:Ambient
Frequently Bought Together