Order Entry
United Kingdom
ContactUsLinkComponent
Nickel(II) sulphate hexahydrate 98%
Toxic
Health
Nickel(II) sulphate hexahydrate 98%
Catalog # APOSIN2713-1KG
Supplier:  Apollo Scientific
CAS Number:  
Toxic
Health
Nickel(II) sulphate hexahydrate 98%
Catalog # APOSIN2713-1KG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :262.85 g/mol
Density:2.07 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3288
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:10101-97-0
Formula:NiSO₄·6H₂O
MDL Number:MFCD00149813