
Gallium 99.9999%, ingot
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7440-55-3
:
Formula:Ga
MDL Number:MFCD00134045
:69.72 g/mol
Boiling Pt:2403 °C/760mmHg
Melting Pt:29.78 °C
Density:5.91 g/cm³ (25 °C)
Storage Temperature:Refrigerator
UN:2803
ADR:8,III
:GHS05
Frequently Bought Together