
Gadolinium trichloride hexahydrate 99,9%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13450-84-5
:
Formula:Cl₃Gd·6H₂O
MDL Number:MFCD00149732
:371.7 g/mol
Density:2.424 g/cm³ (25 °C)
EINECS:233-386-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium dihydrogen phosphate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium iodide ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium chloride ≥98%, TECHNICAL
Supplier: VWR Chemicals
Sodium sulphate, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Copper(II) sulphate pentahydrate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Dysprosium trichloride hydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific