Order Entry
United Kingdom
ContactUsLinkComponent
Erbium 99,9%, chips
Erbium 99,9%, chips
Catalog # APOSIN1678-10G
Supplier:  Apollo Scientific
CAS Number:  
Erbium 99,9%, chips
Catalog # APOSIN1678-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :167.26 g/mol
Boiling Pt:2900 °C
Melting Pt:1497 °C
Density:9.05 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:7440-52-0
Formula:Er
MDL Number:MFCD00010987