Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Copper(II) oxide 99.99%

Copper(II) oxide 99.99%

Irritant
Environment
Supplier:   Apollo Scientific
Copper(II) oxide 99.99%
Supplier:  Apollo Scientific
Certificates
Copper(II) oxide 99.99%

2 Options Available Below

Product Details & Documents
Synonyms:  Cupric oxide, Copper oxide, Copper monoxide granulated
  • CAS Number:1317-38-0
  • Formula:CuO
  • MDL Number:MFCD00010979
  • Molecular Weight:79.55 g/mol
  • Melting Pt:1362 °C
  • Density:6.315 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • UN:3077
  • ADR:9+EHS,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £308.00
    £308.00
    1 * 100 g
       £308.00

  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - £946.00
    £946.00
    1 * 500 g
       £946.00