Copper(I) iodide 99.998%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7681-65-4
:1335-23-5
Formula:CuI
MDL Number:MFCD00010978
:190.45 g/mol
Boiling Pt:1290 °C
Melting Pt:605 °C
Density:5.62 g/cm³ (25 °C)
EINECS:231-674-6
UN:3077
ADR:9+EHS,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together