Cobalt 99.9+%, pieces
Catalog # APOSIN1483-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:58.93 g/mol
Boiling Pt:2900 °C
Melting Pt:1495 °C
Density:8.92 g/cm³ (25 °C)
UN:3077
ADR:9+EHS,III
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:7440-48-4
Formula:Co
MDL Number:MFCD00010935