Order Entry
United Kingdom
ContactUsLinkComponent
Deuterium oxide (>99.9% D)
Irritant
Deuterium oxide (>99.9% D)
Catalog # APOSDE50-10X0.75ML
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Deuterium oxide (>99.9% D)
Catalog # APOSDE50-10X0.75ML
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    10
  • Seq:
    0010
  • Pk Info:
    10× 0.75 ml

Specifications

About this item

 :
 :20 g/mol
EINECS:232-148-9
 :GHS07
Storage Temperature:Ambient
CAS Number:7789-20-0
Formula:D₂O
MDL Number:MFCD00044636