Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Wortmannin
Wortmannin (from Penicillium funiculosum)
Wortmannin (from Penicillium funiculosum)
APOSBIW4201-5MG
:
Apollo Scientific
:
Wortmannin (from Penicillium funiculosum)
APOSBIW4201-5MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 mg
Seq:
2210
:
:
428.44 g/mol
:
234…240 °C
:
Freezer
:
3462
:
6.1,I
:
GHS06
:
19545-26-7
:
C₂₃H₂₄O₈
:
MFCD00133927