Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
AZD-0156
AZD-0156 99%
AZD-0156 99%
AZD-0156 99%
APOSBISN0034-5MG
:
Apollo Scientific
:
AZD-0156 99%
APOSBISN0034-5MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 mg
Seq:
2210
:
:
461.57 g/mol
:
Refrigerator
:
1821428-35-6
:
C₂₆H₃₁N₅O₃
:
MFCD30470661