Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Pterostilbene

Pterostilbene

Corrosive
Environment
Supplier:   Apollo Scientific
Pterostilbene
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  (E)-4-(3,5-Dimethoxystyryl)phenol, 4-[(E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol
  • CAS Number:537-42-8
  • Formula:C₁₆H₁₆O₃
  • MDL Number:MFCD00238710
  • Molecular Weight:256.2964 g/mol
  • Storage Temperature:Refrigerator
  • UN:3077
  • ADR:9+EHS,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £27.50
    £27.50
    1 * 5 g
       £27.50

  • Bottle
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - £51.70
    £51.70
    1 * 10 g
       £51.70

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £110.00
    £110.00
    1 * 25 g
       £110.00