
1,2-Naphthoquinone-4-sulphonic acid sodium salt
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:521-24-4
:
Formula:C₁₀H₅NaO₅S
MDL Number:MFCD00001700
:260.20 g/mol
Melting Pt:289 °C
EINECS:208-308-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together