

Doxorubicin hydrochloride 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:25316-40-9
:
Formula:C₂₇H₂₉NO₁₁·HCl
MDL Number:MFCD00077757
:579.98 g/mol
Melting Pt:216 °C (dec.)
EINECS:246-818-3
:GHS07
:GHS08
Storage Temperature:Refrigerator
Frequently Bought Together