

Cytochalasin D
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22144-77-0
:
Formula:C₃₀H₃₇NO₆
MDL Number:MFCD00077706
:507.63 g/mol
Melting Pt:260…267 °C
Storage Temperature:Freezer
EINECS:244-804-1
UN:1544
ADR:6.1,II
:GHS06
:GHS08
Frequently Bought Together