£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
Resiquimod
Resiquimod
Resiquimod
APOSBI3860-1G
:
Apollo Scientific
:
Resiquimod
APOSBI3860-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
314.389 g/mol
:
166 °C
:
GHS07
:
Freezer
:
144875-48-9
:
C₁₇H₂₂N₄O₂
:
MFCD00937759