Anti-S protein RBD neutralising antibody HEK293 monoclonal antibody (Trehalose free)
ACRBS1N-M13A1-1MG
- Antibody type:Primary
- Antigen name:S protein RBD
- Antigen symbol:S protein RBD
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. No cross-reactivity is deteced with Spike protein RBD domain of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.
- Form:Liquid
- Antigen synonyms:Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19
- Shipping temperature:Dry Ice
- Purification:Affinity purified
- Pk:1 mg
Type: Primary
Antigen: S protein RBD
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: HEK293
Isotype: Human IgG1/kappa
Reactivity: