Order Entry
United Kingdom
ContactUsLinkComponent
6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-amine 95%
6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-amine 95%
Catalog # APOSPC905429-250MG
Supplier:  Apollo Scientific
CAS Number:  
6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-amine 95%
Catalog # APOSPC905429-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :201.149 g/mol
Storage Temperature:Freezer
CAS Number:1005785-87-4
Formula:C₈H₆F₃N₃
MDL Number:MFCD11846602