Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Fluoro-3-nitrophenylacetonitrile
4-Fluoro-3-nitrophenylacetonitrile 95%
4-Fluoro-3-nitrophenylacetonitrile 95%
APOSPC905355-1G
:
Apollo Scientific
:
4-Fluoro-3-nitrophenylacetonitrile 95%
APOSPC905355-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
:
180.136 g/mol
:
Ambient
:
157662-77-6
:
C₈H₅FN₂O₂
:
MFCD09909410