Order Entry
United Kingdom
ContactUsLinkComponent
2,3,5,6-Tetrafluoro-4-hydroxybenzaldehyde 95%
2,3,5,6-Tetrafluoro-4-hydroxybenzaldehyde 95%
Catalog # APOSPC903690-1G
Supplier:  Apollo Scientific
CAS Number:  
2,3,5,6-Tetrafluoro-4-hydroxybenzaldehyde 95%
Catalog # APOSPC903690-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.083 g/mol
Storage Temperature:Ambient
CAS Number:24336-73-0
Formula:C₇H₂F₄O₂
MDL Number:MFCD11505943