1,2,3-Trifluoro-5-methylbenzene 95%
Catalog # APOSPC902729-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:146.11 g/mol
Storage Temperature:Ambient
CAS Number:284463-96-3
Formula:C₇H₅F₃
MDL Number:MFCD03425901