
5-[(4-Chlorobenzyl)thio]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:321553-32-6
:
Formula:C₁₃H₁₀ClF₃N₂OS
MDL Number:MFCD00140838
Melting Pt:108…109 °C
:334.745 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together