1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carbonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:306934-79-2
:
Formula:C₁₁H₁₁ClF₃N₃O
MDL Number:MFCD02091616
:293.673 g/mol
Boiling Pt:158 °C (2 mm Hg)
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together