Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Dimethoxymethyl)-2-fluoroaniline
3-(Dimethoxymethyl)-2-fluoroaniline 95%
3-(Dimethoxymethyl)-2-fluoroaniline 95%
3-(Dimethoxymethyl)-2-fluoroaniline 95%
APOSPC56299-5G
:
Apollo Scientific
:
3-(Dimethoxymethyl)-2-fluoroaniline 95%
APOSPC56299-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
185.195 g/mol
:
Ambient
:
2244087-96-3
:
C₉H₁₂FNO₂
:
MFCD31614287