2-(2,2,3,4,4,4-Hexafluorobutoxy)ethanol
Catalog # APOSPC56226-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-(2,2,3,4,4,4-Hexafluorobutoxy)ethanol
:
:226.117 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:63693-10-7
Formula:C₆H₈F₆O₂
MDL Number:MFCD04972891