Methyl 3-fluoro-2-(trifluoromethyl)isonicotinate > 98%
Catalog # APOSPC540020-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:223.124 g/mol
Storage Temperature:Refrigerator
CAS Number:1227594-82-2
Formula:C₈H₅F₄NO₂
MDL Number:MFCD16611619