Ethyl 2-(4-Chlorophenyl)-2,2-difluoroacetate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:130754-19-7
:
Formula:C₁₀H₉ClF₂O₂
MDL Number:MFCD11007726
:234.627 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together