
4-(Dimethylcarbamoylmethoxy)-3-fluorobenzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1548330-46-6
:
Formula:C₁₀H₁₁ClFNO₄S
MDL Number:MFCD24328309
:295.715 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together