
N-[1-(4-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2379918-45-1
:
Formula:C₁₁H₁₂FN·ClH
MDL Number:MFCD31745722
:213.679 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together