[3-(Chloromethyl)-5-(difluoromethyl)-4-hydroxy-phenyl]azinate
Catalog # APOSPC520306-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:237.588 g/mol
Storage Temperature:Ambient
CAS Number:2092612-17-2
Formula:C₈H₆ClF₂NO₃
MDL Number:MFCD30496625