Order Entry
United Kingdom
ContactUsLinkComponent
2-Bromo-7-(difluoromethyl)-5H-pyrrolo[2,3-b]pyrazine
2-Bromo-7-(difluoromethyl)-5H-pyrrolo[2,3-b]pyrazine
Catalog # APOSPC520169-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-7-(difluoromethyl)-5H-pyrrolo[2,3-b]pyrazine
Catalog # APOSPC520169-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :248.028 g/mol
Storage Temperature:Ambient
CAS Number:2092061-78-2
Formula:C₇H₄BrF₂N₃
MDL Number:MFCD29767649